missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABAT Monoclonal Antibody (OTI6H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806898S
This item is not returnable.
View return policy
Description
ABAT Monoclonal antibody specifically detects ABAT in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ABAT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P80404 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 18 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6H1 | |
| Unconjugated | |
| ABAT | |
| (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase; 4-aminobutyrate aminotransferase, brain isoform; 4-aminobutyrate aminotransferase, liver isoform; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; 9630038C02Rik; ABAT; AI255750; beta-AlaAT; beta-alanine oxoglutarate aminotransferase; cb880; ENSMUSG00000051226; fj82a01; FLJ17813; FLJ30272; GABA aminotransferase; GABA AT; GABA T; GABA transaminase; GABA transferase; Gabaat; GABA-AT; Gabat; GABA-T; Gamma-amino-N-butyrate transaminase; Gm9851; hCG1984265; I54; L AIBAT; Laibat; L-AIBAT; mitochondrial 4-aminobutyrate aminotransferase; NPD009; wu:fj82a01; X61497 | |
| Human recombinant protein fragment corresponding to amino acids 29-323 of human ABAT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction