missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRPB2 Antibody (1008709) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9998JF525
This item is not returnable.
View return policy
Description
SIRPB2 Monoclonal antibody specifically detects SIRPB2 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| SIRPB2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG | |
| HEK293 human embryonic kidney cell line, HEK293 derived human SIRP beta 2, Gln33-Gly287, Accession # Q5JXA9 | |
| RUO | |
| 284759 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 1008709 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction