missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIRT1 Monoclonal Antibody (3H10A10)
GREENER_CHOICE

Product Code. 16363604
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16363604 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16363604 Supplier Invitrogen™ Supplier No. MA529570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SIRT1.

NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metabolism, apoptosis, and autophagy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5'-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT1
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 3H10A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SIRT1
Gene Accession No. Q96EB6
Gene Alias 75SirT1; AA673258; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; SIR2; Sir2a; SIR2alpha; SIR2L1; sir2-like 1; SIR2-like protein 1; SIRT 1; SIRT1; SIRT-1; SirtT1 75 kDa fragment; sirtuin 1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1
Gene Symbols SIRT1
Host Species Mouse
Immunogen Recombinant Human SIRT1 protein (Met193-Ser747).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.