missing translation for 'onlineSavingsMsg'
Learn More

SIRT1 Rabbit, anti-Human, Clone: 2F11, Proteintech

Product Code. p-200050176 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024505 100 μL 100µL
18054275 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024505 Supplier Proteintech Supplier No. 827904RR100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The SIRT1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SIRT1 . This antibody recognizes human antigen. The SIRT1 antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Antigen SIRT1
Applications Western Blot, Immunofluorescence, ELISA
Clone 2F11
Concentration 400 μg/mL
Gene Symbols hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.