Learn More
SIRT1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL750-13161-20UL
 View more versions of this product
Product Code. 18030526
  • € 140.00 / 20µL
Description
The SIRT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIRT1. This antibody recognizes human antigen. The SIRT1 antibody has been validated for the following applications: FC (Intra), WB analysis. This antibody is coralite™ plus 750.
Specifications
Specifications
SIRT1
1000 μg/mL
hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1
20 μL
23411
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Western Blot, Flow Cytometry (Intracellular Staining)
CoraLite Plus 750
Rabbit
Primary
Human
150 kDa