Learn More
SIRT1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL750-13161-100UL
Description
The SIRT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIRT1. This antibody recognizes human antigen. The SIRT1 antibody has been validated for the following applications: FC (Intra), WB analysis. This antibody is coralite™ plus 750.
Specifications
| SIRT1 | |
| 1000 μg/mL | |
| hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1 | |
| 100 μL | |
| 23411 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 750 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |