missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT2, Mouse anti-Human, Clone: S2R233-1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16061.100ug
This item is not returnable.
View return policy
Specifications
| SIRT2 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4. | |
| SIR2/SIR2L/SIR2L2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| S2R233-1 | |
| ELISA (1:2000-5000) Western Blot (1:2000-5000) The optimal working dilution should be determined by the end user. | |
| sirtuin (silent mating type information regulation 2 homolog) 2 (S. cerevisiae) | |
| SIRT2 | |
| Recombinant protein corresponding to human SIRT2. | |
| RUO | |
| 22933 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction