Learn More
SIRT2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D8G10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66410-1-IG-20UL
 View more versions of this product
Product Code. 16818265
  • € 145.00 / 20µL
Description
The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.
Specifications
Specifications
SIRT2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5RJQ4, Q8IXJ6, Q8VDQ8
Sirt2
SIRT2 Fusion Protein Ag7756
20 μL
Primary
Human, Rat, Pig, Mouse
Antibody
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
1D8G10
Unconjugated
SIRT2
SIR2, SIR2 like protein 2, SIR2L, SIR2L2, SIRT2
Mouse
Protein G
RUO
100125964, 22933, 361532, 64383
-20°C