missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRT3 Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550864
This item is not returnable.
View return policy
Description
SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The SIRT3 is included in class I of the sirtuin family.
Specifications
| SIRT3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q9NTG7 | |
| SIRT3 | |
| Recombinant Human SIRT3 protein, Gln126-Lys399 (Accession #Q9NTG7). | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| C3 | |
| Unconjugated | |
| SIRT3 | |
| 2310003L23Rik; AI848213; HGNC:14931; hSIRT3; mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase; mitochondrial protein lysine deacetylase; mSIR2L3; NAD-dependent deacetylase sirtuin-3; NAD-dependent deacetylase sirtuin-3, mitochondrial; NAD-dependent protein deacetylase sirtuin-3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; regulatory protein SIR2 homolog 3; silent mating type information regulation 2, (S.cerevisiae, homolog)-like 3; silent mating type information regulation 2, S.cerevisiae, homolog 3; Sir2l3; sir2-like 3; SIR2-like protein 3; SIRT3; SIRT3L mitochondrial; sirtuin 3; sirtuin type 3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100125971, 23410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction