missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT6 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2524AF532
This item is not returnable.
View return policy
Description
SIRT6 Polyclonal specifically detects SIRT6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SIRT6 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
| Rabbit | |
| 0.1 mL | |
| 51548 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SIRT6 | |
| A synthetic peptide made to a C-terminal region of the human SIRT6 protein (within residues 300-355). [Swiss-Prot Q8N6T7] | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction