missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT6 Mouse anti-Human, Clone:2G1H1, Abnova™
Mouse monoclonal antibody raised against recombinant human SIRT6.
Brand: Abnova MAB17126.100ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family. [provided by RefSeq]Specifications
| SIRT6 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| SIRT6 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry, Western Blot | |
| 2G1H1 | |
| sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae) | |
| In PBS (0.05% sodium azide) | |
| SIR2L6 | |
| Recombinant protein corresponding to amino acid 141-250 of human SIRT6 from E. coli. | |
| 51548 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction