missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12994-1-AP-150UL
This item is not returnable.
View return policy
Description
SIRT7 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. SIRT7 is included in class IV. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. SIRT7 is included in class IV of the sirtuin family.Specifications
| SIRT7 | |
| Polyclonal | |
| Unconjugated | |
| SIRT7 | |
| SIR2 like protein 7, SIR2L7, SIRT7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 209011, 303745, 51547 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B2RZ55, Q8BKJ9, Q9NRC8 | |
| Sirt7 | |
| SIRT7 Fusion Protein Ag3656 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction