missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB12032.100uL
This item is not returnable.
View return policy
Specifications
| SIRT7 | |
| Polyclonal | |
| Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae) | |
| SIRT7 | |
| A synthetic peptide corresponding to amino acids 100-200 of human SIRT7. | |
| RUO | |
| 51547 | |
| Store at 4°C. Do not freeze. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| MGC126840/MGC126842/SIR2L7 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction