missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody (012), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89977H
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for ELISA
Specifications
| Sirtuin 1/SIRT1 | |
| Monoclonal | |
| HRP | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Sirtuin 1/SIRT1 (NP_036370.2; Met 193-Ser 747). | |
| 0.1 mL | |
| Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific | |
| 23411 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 012 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction