missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 1/SIRT1, p Thr530 Antibody (SR1099), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21557-100ul
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot
Specifications
| Sirtuin 1/SIRT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1099 | |
| Western Blot 1:1000-1:2000 | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| A synthesized peptide derived from human phospho-Sirtuin 1/SIRT1 [Thr530] (Uniprot #: Q96EB6) | |
| 100 μg | |
| Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific | |
| 23411 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction