missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 2/SIRT2 Antibody (PCRP-SIRT2-1A8), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08293MFV450
This item is not returnable.
View return policy
Description
Sirtuin 2/SIRT2 Monoclonal antibody specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Flow Cytometry
Specifications
| Sirtuin 2/SIRT2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-SIRT2-1A8 | |
| Flow Cytometry | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
| Recombinant full-length human Sirtuin 2/SIRT2 protein (Uniprot: Q8IXJ6 ) | |
| 0.1 mL | |
| Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
| 22933 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction