missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 3/SIRT3 Antibody (6C12D1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52562
This item is not returnable.
View return policy
Description
Sirtuin 3/SIRT3 Monoclonal specifically detects Sirtuin 3/SIRT3 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
| Sirtuin 3/SIRT3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3 | |
| Mouse | |
| 43.6 kDa | |
| 0.1 mg | |
| Epigenetics, Histone Deacetylases | |
| 23410 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| 6C12D1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SIRT3 | |
| Purified recombinant fragment of human SIRT3 (AA: 155-290) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction