missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 3/SIRT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 235.00 - € 478.80
Specifications
| Antigen | Sirtuin 3/SIRT3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18377196
|
Novus Biologicals
NBP3-02984-20UL |
20 μL |
€ 235.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18025244
|
Novus Biologicals
NBP3-02984-100UL |
100 μL |
€ 507.00 € 478.80 / 100µL Save € 28.20 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sirtuin 3/SIRT3 Polyclonal specifically detects Sirtuin 3/SIRT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Sirtuin 3/SIRT3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human Sirtuin 3/SIRT3 (NP_036371.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS with 50% glycerol, pH7.3. | |
| 23410 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title