Learn More
Sirtuin 5/SIRT5 Antibody (9H12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26323-100ul
 View more versions of this product
Product Code. 30031783
  • € 533.00 / 100µL
Description
Sirtuin 5/SIRT5 Monoclonal antibody specifically detects Sirtuin 5/SIRT5 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sirtuin 5/SIRT5
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
9H12
ELISA, Immunohistochemistry 1:50-1:200
EC 3.5.1, EC 3.5.1.-, FLJ36950, NAD-dependent deacetylase sirtuin-5, silent mating type information regulation 2, S.cerevisiae, homolog 5, SIR2L5, sir2-like 5, SIR2-like protein 5, sirtuin (silent mating type information regulation 2 homolog) 5 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 5, sirtuin 5, sirtuin type 5
A synthesized peptide derived from Human Sirtuin 5/SIRT5 [UniProt Q9NXA8]
100 μL
Epigenetics
23408
Store at -20 to -70C. Avoid freeze-thaw cycles.