missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sitafloxacin hydrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Sitafloxacin (DU6859a) hydrate is a potent, orally active fluoroquinolone antibiotic with in vitro activity against a broad range of gram-positive and gram-negative bacteria, including anaerobic bacteria, as well as against atypical pathogens. Sitafloxacin can be used for the research of respiratory tract infection and urinary tract infection.
Specifications
Specifications
| Chemical Name or Material | Sitafloxacin hydrate |
| Physical Form | Solid |
| CAS | 163253-35-8 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18ClF2N3O3.3/2H2O |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | DU6859a hydrate |
| Solubility Information | DMSO : 8.9 mg/mL (20.37 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (2.29 mM; ultrasonic and warming and heat to 80°C) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction