missing translation for 'onlineSavingsMsg'
Learn More

Sitaxsentan sodium, MedChemExpress

Product Code. p-200095923
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495286

Brand: Medchem Express HY11103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sitaxsentan sodium (IPI 1040 sodium; TBC11251 sodium) is an orally active, highly selective antagonist of endothelin A receptors.

Sitaxsentan sodium (IPI 1040 sodium

Specifications

Chemical Name or Material Sitaxsentan sodium
CAS 210421-74-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H14ClN2NaO6S2
Quantity 100 mg
Synonym IPI 1040 sodiumTBC11251 sodium
Solubility Information DMSO : 100 mg/mL (209.69 mM; Need ultrasonic) ∣H2O : 100 mg/mL (209.69 mM; Need ultrasonic)
SMILES O=S(C1=C(SC=C1)C(CC2=C(C=C3OCOC3=C2)C)=O)(N([Na])C4=C(C(C)=NO4)Cl)=O
Molecular Weight (g/mol) 476.89
Formula Weight 476.89
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.