missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIVA Antibody (OTI2F8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74208CL1
This item is not returnable.
View return policy
Description
SIVA Monoclonal antibody specifically detects SIVA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SIVA | |
| Monoclonal | |
| CoraFluor 1 | |
| CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
| Full length human recombinant protein of human SIVA1(NP_006418) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis | |
| 10572 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2F8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction