missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIVA1 Monoclonal Antibody (OTI1C9)
GREENER_CHOICE

Product Code. 15945874
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15945874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15945874 Supplier Invitrogen™ Supplier No. MA526277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIVA1 Monoclonal Antibody for Western Blot, ICC/IF

Induces CD27-mediated apoptosis. Inhibits BCL2L1 isoform Bcl-x(L) anti-apoptotic activity. Inhibits activation of NF-kappa-B and promotes T-cell receptor-mediated apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIVA1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SIVA1
Gene Accession No. O15304
Gene Alias apoptosis regulatory protein Siva; Cd27 binding protein; CD27-binding (Siva) protein; CD27-binding protein; CD27BP; SIVA; SIVA1; Siva-1; SIVA1 apoptosis inducing factor; SIVA1, apoptosis-inducing factor; Siva-2
Gene Symbols SIVA1
Host Species Mouse
Immunogen Full length human recombinant protein of SIVA1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10572
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.