missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIVA1 Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506828S
This item is not returnable.
View return policy
Description
SIVA1 Monoclonal antibody specifically detects SIVA1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SIVA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15304 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10572 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| SIVA1 | |
| apoptosis regulatory protein Siva; Cd27 binding protein; CD27-binding (Siva) protein; CD27-binding protein; CD27BP; SIVA; SIVA1; Siva-1; SIVA1 apoptosis inducing factor; SIVA1, apoptosis-inducing factor; Siva-2 | |
| Full length human recombinant protein of human SIVA1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction