missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ Six Block Digital Dry Block Heater HB6DG
1626.35 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Use promo code "21657" to get your promotional price.
Click to view available options
Description:
Dry Block Heater, 6 Block, HB6DG, EU
Description
- Designed for applications that require repeatable results and superior temperature stability
- These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
- Integral support rod holder with locking knob accepts optional external temperature probe kit
- Optional external temperature probe kit monitors actual block or sample temperature
- Each of the five models accepts separate interchangeable modular blocks (available separately) accommodating various tube sizes from 0.2 mL microtubes to 50 mL centrifuge tubes
- Each block has a thermometer well for measuring block temperature
- Efficient heating due to close tube-and-block contact for maximum heat retention
Specifications
Certifications/Compliance | TUV |
Controller Type | Digital |
Description | Dry Block Heater, 6 Block, HB6DG, EU |
Temperature Range (Metric) | 5°C to 120°C |
Type | Dry Bath Block |
Heat Up Time | 65 min |
Model | HB6DG |
No. of Blocks | 6 |
Temperature Stability | ±2°C |
Temperature Uniformity | ±0.1°C at 37°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'