missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIX1 Mouse anti-Human, Clone: OTI1H4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504057
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a homeobox protein that is similar to the Drosophila ′sine oculis′ gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal domit deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).Specifications
| SIX1 | |
| Monoclonal | |
| 0.77 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15475 | |
| SIX1 | |
| Human recombit protein fragment corresponding to amino acids 1-284 of human SIX1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1H4 | |
| Unconjugated | |
| SIX1 | |
| BOS3, DFNA23, TIP39 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6495 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction