missing translation for 'onlineSavingsMsg'
Learn More

(+)-SJ733, MedChemExpress

Product Code. 16490268
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490268 5 mg 5mg
16471829 1 mg 1mg
16457808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490268 Supplier Medchem Express Supplier No. HY19556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (+)-SJ733 is an anti-malaria agent which can also inhibit Na+-ATPase PfATP4.

(+)-SJ733 is an anti-malaria agent which can also inhibit Na+-ATPase PfATP4.

Specifications

Chemical Name or Material (+)-SJ733
Physical Form Solid
CAS 1424799-20-1
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H16F4N4O2
For Use With (Application) COVID-19-immunoregulation
Synonym SJ000557733
Solubility Information DMSO : 50 mg/mL (106.75 mM; Need ultrasonic)
SMILES O=C([C@@H](C1=C2C=CC=C1)[C@@H](C3=CC=CN=C3)N(CC(F)(F)F)C2=O)NC4=CC=C(F)C(C#N)=C4
Molecular Weight (g/mol) 468.4
Formula Weight 468.4
Percent Purity 99.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.