missing translation for 'onlineSavingsMsg'
Learn More

SJB2-043, MedChemExpress

Product Code. 16469347
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469347 5 mg 5mg
16486798 10 mg 10mg
16476916 100 mg 100mg
16412877 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469347 Supplier Medchem Express Supplier No. HY15757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SJB2-043 is an inhibitor of the native USP1/UAF1 complex with IC50 of 544 nM.

SJB2-043 is an inhibitor of the native USP1/UAF1 complex with IC50 of 544 nM.

Specifications

Chemical Name or Material SJB2-043
Physical Form Solid
CAS 63388-44-3
Color Deep Green
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H9NO3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 3.33 mg/mL (12.10 mM; Need ultrasonic)
SMILES O=C1C2=C(N=C(C3=CC=CC=C3)O2)C(C4=CC=CC=C41)=O
Molecular Weight (g/mol) 275.26
Formula Weight 275.26
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.