Learn More
Tocris Bioscience™ (+)-SK&F 10047 hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
(+)-SK&F 10047 hydrochloride is a prototypical σ1 receptor agonist. Behaviorally active in animal models of memory and stress.
Specifications
Specifications
| Chemical Name or Material | (2S,6S,11S)-1,2,3,4,5,6-Hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol hydrochloride |
| CAS | 133005-41-1 |
| Quantity | 50 mg |
| Target | Sigma1 Receptor Agonists |
| Molecular Formula | C17H23NO.HCl |
| Purity | 0.98 |