missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SKA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585456
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (94%), Bovine (87%). Positive Control: Target recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Upon entry into mitosis, the cell's microtubule (MT) network forms the mitotic spindle, allowing the segregation of paired chromosomes. Proteinaceous structures on centromeric chromatin termed kinetochores (KT) are essential for the proper attachment of the chromosomes to the spindle MTs. A recently discovered spindle and kinetochore complex, comprised of proteins SKA1, SKA2, and SKA3, has been found to be required for stable KT-MT interactions and timely anaphase onset. Depletion of either SKA1 or SKA2 by siRNA results in the loss of both proteins from the KT, but does not impact overall KT structure. Cells depleted of the SKA complex undergo a prolonged checkpoint-dependent delay in a metaphase-like state, indicating the importance of the SKA complex in the maintenance of the metaphase plate and spindle checkpoint silencing.
Specifications
| SKA1 | |
| Polyclonal | |
| Unconjugated | |
| SKA1 | |
| 2810433K01Rik; AV117428; C18orf24; RGD1310784; SKA1; spindle and kinetochore associated complex subunit 1; spindle and kinetochore-associated protein 1; spindle and KT (kinetochore) associated 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 220134, 291441, 66468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 20% glycerol, 1% BSA and 0.01% thimerosal, pH 7 | |
| B0BN28, Q96BD8, Q9CPV1 | |
| SKA1 | |
| Recombinant protein encompassing a sequence within the center region of human SKA1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction