missing translation for 'onlineSavingsMsg'
Learn More

SKF-86002, MedChemExpress

Product Code. 16465228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465228

Brand: Medchem Express HY12511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKF-86002 is an orally active p38 MAPK inhibitor, with anti-inflammatory, anti-arthritic and analgesic activities. SKF-86002 inhibits lipopolysaccharide (LPS)-stimulate human monocyte IL-1 and TNF-α production (IC50 = 1 μM). SKF-86002 inhibits lipoxygenase- and cyclooxygenase-mediated metabolism of arachidonic acid.

SKF-86002 is an orally active p38 MAPK inhibitor, with anti-inflammatory, anti-arthritic and analgesic activities. SKF-86002 inhibits lipopolysaccharide (LPS)-stimulate human monocyte IL-1 and TNF-α production (IC50 = 1 μM). SKF-86002 inhibits lipoxygenase- and cyclooxygenase-mediated metabolism of arachidonic acid.

Specifications

Chemical Name or Material SKF-86002
CAS 72873-74-6
Color Brown
Purity Grade Notes Research
Molecular Formula C16H12FN3S
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (112.09 mM; Need ultrasonic)
SMILES FC1=CC=C(C2=C(C3=CC=NC=C3)N4C(SCC4)=N2)C=C1
Molecular Weight (g/mol) 297.35
Formula Weight 297.35
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.