missing translation for 'onlineSavingsMsg'
Learn More

SKI Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18659551 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18659551 0.1 mL 0.1mL
18628022 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18659551 Supplier Novus Biologicals Supplier No. NBP2941050.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SKI Polyclonal antibody specifically detects SKI in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKI
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias C-ski, Proto-oncogene c-Ski, ski oncogene, ski oncoprotein, SKV, v-ski avian sarcoma viral oncogene homolog, v-ski sarcoma viral oncogene homolog (avian)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 650-728 of human SKI (NP_003027.1). CDKGCEAGRLRAKYSAQIEDLQVKLQHAEADREQLRADLLREREAREHLEKVVKELQEQLWPRARPEAAGSEGAAELEP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6497
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.