missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Antibody (D1L165-6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80048DL594
This item is not returnable.
View return policy
Description
DLL1 Monoclonal antibody specifically detects DLL1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| DLL1 | |
| Monoclonal | |
| DyLight 594 | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Recombinant human DLL1 | |
| 0.1 mL | |
| Neuroscience, Signal Transduction, Stem Cell Signaling Pathway | |
| 28514 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| D1L165-6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction