Learn More
Invitrogen™ SKIV2L Monoclonal Antibody (K94011_4C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556090
Description
Sequence of this protein is as follows: SEFPSRKDSK AHEQALAELT KRLGALEEPD MTGQLVDLPE YYSWGEELTE TQHMIQRRIM ESVNGLKSLS AGRVVVVKNQ EHHNALGVIL QVSSNSTSRV FTTLVLCDKP LSQDPQDRGP ATAEVPYPDD LVGFKLFLPE GPCDHTVVKL QPGDMAAITT KVLRVNGEKI LEDFSKRQQP KFKKDPPLAA VTTAVQELLR LAQAHPAGPP TLDPVNDLQL KDMSVVEGGL RARKLEELIQ GAQCVHSPRF PAQYLKLRER MQIQKEMERL
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.
Specifications
| SKIV2L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q15477 | |
| SKIV2L | |
| Recombinant human SKIC2 (immunogen range is 771-1040). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| K94011_4C6 | |
| Unconjugated | |
| SKIV2L | |
| 170A; 4930534J06Rik; AW214248; Ddx13; Helicase SKI2W; helicase-like protein; HLP; SKI; SKI2; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; Ski2w; SKIV2; SKIV2L; SKIV2L1; superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like (S. cerevisiae); THES2; W | |
| Mouse | |
| Protein A | |
| RUO | |
| 6499 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.