missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKIV2L2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | SKIV2L2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100 - 1:500, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672622
|
Novus Biologicals
NBP2-94315-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606012
|
Novus Biologicals
NBP2-94315-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SKIV2L2 Polyclonal antibody specifically detects SKIV2L2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| SKIV2L2 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ATP-dependent helicase SKIV2L2, Dob1, EC 3.6.1, EC 3.6.4.13, fSAP118, KIAA0052functional spliceosome-associated protein 118, MGC142069, Mtr4superkiller viralicidic activity 2-like 2, superkiller viralicidic activity 2-like 2 (S. cerevisiae) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-304 of human SKIV2L2 (NP_056175.3). MADAFGDELFSVFEGDSTTAAGTKKDKEKDKGKWKGPPGSADKAGKRFDGKLQSESTNNGKNKRDVDFEGTDEPIFGKKPRIEESITEDLSLADLMPRVKVQSVETVEGCTHEVALPAEEDYLPLKPRVGKAAKEYPFILDAFQREAIQCVDNNQSVLVSAHTSAGKTVCAEYAIALALREKQRVIFTSPIKALSNQKYREMYEEFQDVGLMTGDVTINPTASCLVMTTEILRSMLYRGSEVMREVAWVIFDEIHYMRDSERGVVWEETIILLPDNVHYVFLSATIPNARQFAEWICHLHKQPC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100 - 1:500, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 23517 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title