missing translation for 'onlineSavingsMsg'
Learn More

SKLB4771, MedChemExpress

Product Code. 16461627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461627

Brand: Medchem Express HY12960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKLB4771 is a novel potent and selective Flt3 inhibitor with IC50 of 10 nM; against FLT3-ITD-expressing MV4-11 cells with IC50 of 6 nM.

SKLB4771 is a novel potent and selective Flt3 inhibitor with IC50 of 10 nM

Specifications

Chemical Name or Material SKLB4771
CAS 1370256-78-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H27N7O3S2
Quantity 50 mg
Synonym FLT3-​IN-​1
Solubility Information DMSO : 33.33 mg/mL (61.99 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(SC2=C3C=CC(OCCCN4CCOCC4)=CC3=NC=N2)S1)NC5=CC=C(C)C=C5
Molecular Weight (g/mol) 537.66
Formula Weight 537.66
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.