missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97674IR
This item is not returnable.
View return policy
Description
Skp2 Polyclonal specifically detects Skp2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Skp2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| CDK2/cyclin A-associated protein p45, cyclin A/CDK2-associated protein p45, Cyclin-A/CDK2-associated protein p45, FBL1, F-box protein Skp2, F-box/LRR-repeat protein 1, FBXL1MGC1366, FLB1, p45skp2, S-phase kinase-associated protein 2, S-phase kinase-associated protein 2 (p45) | |
| Produced in rabbits immunized with E. coli-derived Human Skp2 fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6502 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction