missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLA2 Antibody (Z30P1F12*F4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50293AF350
This item is not returnable.
View return policy
Description
SLA2 Monoclonal specifically detects SLA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| SLA2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SLA2 | |
| Peptide Sequence - LRESLSFY | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Z30P1F12*F4 | |
| Western Blot, ELISA, Immunohistochemistry | |
| C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2 | |
| Mouse | |
| Protein A purified | |
| Immunology, Signal Transduction | |
| 84174 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction