missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81827
This item is not returnable.
View return policy
Description
SLAIN1 Polyclonal specifically detects SLAIN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLAIN1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| SLAIN1 | |
| SLAIN1 antibody was raised against a 16 amino acid synthetic peptide near the center of human SLAIN1. The immunogen is located within amino acids 370 - 420 of SLAIN1. Amino Acid Squence: AQTPDQQPNRTNGDK | |
| 0.1 mg | |
| Primary | |
| At least three isoforms of SLAIN1 are known to exist; this antibody will detect only the largest isoform. SLAIN1 antibody is predicted not to cross-react with SLAIN2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin | |
| SLAIN motif family, member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 122060 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction