missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81827IR
This item is not returnable.
View return policy
Description
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLAIN1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 122060 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| SLAIN motif family, member 1 | |
| SLAIN1 antibody was raised against a 16 amino acid synthetic peptide near the center of human SLAIN1. The immunogen is located within amino acids 370 - 420 of SLAIN1. Amino Acid Squence: AQTPDQQPNRTNGDK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction