missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81828F
This item is not returnable.
View return policy
Description
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
Specifications
| SLAIN1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 122060 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| FITC | |
| SLAIN motif family, member 1 | |
| SLAIN1 antibody was raised against an 15 amino acid synthetic peptide near the carboxy terminus of human SLAIN1. The immunogen is located within the last 50 amino acids of SLAIN1. Amino Acid Squence: NGSNLPRSKIAQPVRS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction