Learn More
SLAIN1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81827MFV450
This item is not returnable.
View return policy
Description
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLAIN1 | |
| Rabbit | |
| Primary | |
| Store at 4C in the dark. |
| 50mM Sodium Borate | |
| SLAIN1 antibody was raised against a 16 amino acid synthetic peptide near the center of human SLAIN1. The immunogen is located within amino acids 370 - 420 of SLAIN1. Amino Acid Squence: AQTPDQQPNRTNGDK | |
| 122060 | |
| IgG |