Learn More
Invitrogen™ SLAIN1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598761
Product Code. 16561582
  • € 473.00 / 100µg
Description
SLAIN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
SLAIN1 (SLAIN Motif Family Member 1) is a Protein Coding gene. An important paralog of this gene is SLAIN2. Microtubule plus-end tracking protein that might be involved in the regulation of cytoplasmic microtubule dynamics, microtubule organization and microtubule elongation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLAIN1
Polyclonal
Unconjugated
SLAIN1
9630044O09Rik; AA675320; AW742596; C13orf32; RGD1308626; SLAIN motif family member 1; SLAIN motif family, member 1; SLAIN motif-containing protein 1; SLAIN1
Rabbit
Protein G
RUO
122060
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
4.23 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q8ND83
SLAIN1
Recombinant Human SLAIN motif-containing protein 1 protein (43-136AA).