missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slain2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41117B
This item is not returnable.
View return policy
Description
Slain2 Polyclonal antibody specifically detects Slain2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Slain2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 57606 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| KIAA1458FLJ21611, SLAIN motif family, member 2, SLAIN motif-containing protein 2 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human SLAIN2. The immunogen is located within amino acids 70 - 120 of SLAIN2. Amino Acid Squence: PRRTSSEELRDATS | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction