missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 Antibody (MM0553-6L6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11907AF750
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot.
Specifications
| SLAM/CD150 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SLAMF1 | |
| Human recombinant SLAM/CD150 (extracellular domain) | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot | |
| MM0553-6L6 | |
| Western Blot | |
| CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
| Mouse | |
| 0.1 mL | |
| 6504 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction