missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLAMF8 Recombinant Rabbit Monoclonal Antibody (210)
GREENER_CHOICE

Product Code. 16324704
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16324704 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16324704 Supplier Invitrogen™ Supplier No. MA531249

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse SLAMF8.

This gene encodes a member of the CD2 family of cell surface proteins involved in lymphocyte activation. These proteins are characterized by Ig domains. This protein is expressed in lymphoid tissues, and studies of a similar protein in mouse suggest that it may function during B cell lineage commitment. The gene is found in a region of chromosome 1 containing many CD2 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLAMF8
Applications ELISA
Classification Recombinant Monoclonal
Clone 210
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Slamf8
Gene Accession No. Q9D3G2
Gene Alias 5830408F06Rik; B lymphocyte activator macrophage expressed; BCM-like membrane protein; Blame; B-lymphocyte activator macrophage expressed; CD353; FLJ20442; MGC129578; SBBI42; SLAM family member 8; SLAMF8
Gene Symbols Slamf8
Host Species Rabbit
Immunogen Recombinant Mouse SLAMF8 protein (Met1-Asp231).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74748
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.