missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC10A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555987
This item is not returnable.
View return policy
Description
Immunogen sequence: LPPNCKRTVC LETGSQNVQL CTAILKLAFP PQFIGSMYMF PLLYALFQSA EAGIFVLIYK MYGSEMLHKR DPLDEDEDTD ISYKKLKEEE MADTSYGTVK AENIIMMETA QTS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Transporter for bile acids.
Specifications
| SLC10A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC10A4 | |
| bile acid transporter SLC10A4; E130304D01; Na(+)/bile acid cotransporter 4; P4; SLC10A4; Sodium/bile acid cotransporter 4; solute carrier family 10 (sodium/bile acid cotransporter family), member 4; solute carrier family 10 member 4; solute carrier family 10, member 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EP9 | |
| SLC10A4 | |
| Recombinant protein corresponding to Human SLC10A4. Recombinant protein control fragment (Product #RP-90673). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction