missing translation for 'onlineSavingsMsg'
Learn More

SLC10A7 Polyclonal Antibody, Invitrogen™

Product Code. 15536361
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15536361 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15536361 Supplier Invitrogen Supplier No. PA539465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC10A7 (solute carrier family 10 (sodium/bile acid cotransporter family), member 7), also known as P7, is a 358 amino acid multi-pass membrane protein belonging to the sodium:bile acid symporter family. Existing as seven alternatively spliced isoforms, SLC10A7 is expressed at high levels in liver and lung, moderate levels in placenta, kidney, spleen, and thymus, and low levels in heart, prostate, and testis. A few members of the sodium:bile acid symporter family, such as NTCP (also known as SLC10A1) and Asbt (also known as SLC10A2), are involved in maintaining enterohepatic circulation of bile acids by mediating the first step of active bile transport through membrane barriers of liver and intestine. Other family members, including SLC10A6, play an important role in the cellular delivery of specific prohormones in testis, placenta, adrenal gland and other peripheral tissues. Orphan carriers such as SLC10A7 are uncharacterized and their functions unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC10A7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC10A7
Gene Accession No. Q0GE19, Q5PT53, Q5PT50
Gene Alias Sodium/bile acid cotransporter 7, Na(+)/bile acid cotransporter 7, Solute carrier family 10 member 7, C4orf13, P7, PSEC0051
Gene Symbols SLC10A7
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human SLC10A7
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291942, 76775, 84068
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.