missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC11A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534195
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%) Galago, Rat, Hamster, Bovine, Horse (94%).
This gene encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| SLC11A2 | |
| Polyclonal | |
| Unconjugated | |
| Slc11a2 | |
| AHMIO1; DCT1; Dct-1; Divalent cation transporter 1; divalent metal transporter 1; DMT1; DMT-1; microcytic anemia, viable anaemia; mk; Natural resistance-associated macrophage protein 2; NRAMP 2; Nramp2; OK/SW-cl.20; SLC11A2; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2; solute carrier family 11 member 2; Solute carrier family 11 member 2 (natural resistance-associated macrophage protein 2); van | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.9 mg/mL | |
| PBS with 0.1% sodium azide | |
| P49281 | |
| Slc11a2 | |
| Synthetic 16 amino acid peptide from internal region of human SLC11A2. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction