missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc12a5 Mouse anti-Human, Rat, HRP, Clone:S1-12, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Slc12a5.
Brand: Abnova MAB16983.100ug
This item is not returnable.
View return policy
Specifications
| Slc12a5 | |
| Monoclonal | |
| HRP | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:300) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Slc12a5 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| S1-12 | |
| solute carrier family 12 (potassium-chloride transporter), member 5 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Kcc2 | |
| Recombinant protein corresponding to amino acids 932-1043 of rat Slc12a5. | |
| 171373 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction